siRNA Details
| Gene Symbol: UBXN7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015562.1 | NP_056377.1 | 9 | 923 | ||
| XM_011512670.2 | XP_011510972.1 | 9 | 871 | |||
| XM_011512671.2 | XP_011510973.1 | 8 | 728 | |||
| GenBank mRNA | AB018337.1 | - | 9 | 851 | ||
| AK026451.1 | - | 9 | 876 | |||
| AK303630.1 | - | 8 | 728 | |||
| AK307899.1 | - | 5 | 484 | |||
| BC028986.1 | - | 8 | 755 | |||
| BC047420.1 | - | 8, 9 | 873 | |||

UBXN7
Genomic Map