siRNA Details
| Gene Symbol: HACL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001284413.1 | NP_001271342.1 | 10 | 1268 | ||
| NM_001284415.1 | NP_001271344.1 | 10 | 1271 | |||
| NM_001284416.1 | NP_001271345.1 | 8 | 1103 | |||
| NM_012260.3 | NP_036392.2 | 11 | 1349 | |||
| NR_104315.1 | 9 | 1170 | ||||
| GenBank mRNA | AF161397.1 | - | 11 | 969 | ||
| AJ131753.1 | - | 11 | 1059 | |||
| AK056977.1 | - | 3 | 343 | |||
| AK301546.1 | - | 10 | 985 | |||
| AK301990.1 | - | 8 | 838 | |||
| AK302086.1 | - | 10 | 988 | |||
| BC001627.1 | - | 11 | 1067 | 3538159 | ||
| BC007440.2 | - | 9 | 875 | |||

HACL1
Genomic Map