siRNA Details
| Gene Symbol: DECR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020664.3 | NP_065715.1 | 9 | 1558 | ||
| GenBank mRNA | AJ293009.1 | - | 9 | 1473 | ||
| AK094340.1 | - | 4 | 2042 | |||
| AK225961.1 | - | 9 | 1727 | |||
| AL442076.1 | - | 2 | 685 | |||
| BC010740.1 | - | 9 | 1544 | 3877040 | ||
| BC011968.1 | - | 9 | 1463 | 3866503 | ||

DECR2
Genomic Map