siRNA Details
| Gene Symbol: EDRF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001202438.1 | NP_001189367.1 | 4 | 756 | ||
| NM_015608.2 | NP_056423.2 | 4 | 756 | |||
| NR_110857.1 | 4 | 756 | ||||
| NR_110858.1 | 4 | 756 | ||||
| NR_110859.1 | 4 | 756 | ||||
| XM_017016066.1 | XP_016871555.1 | 4 | 541 | |||
| GenBank mRNA | AK091507.1 | - | 4 | 568 | ||
| BC026172.1 | - | 4 | 757 | 4838438 | ||
| BC105929.1 | - | 4 | 695 | |||
| BC107479.1 | - | 4 | 695 | 40028896 | ||

EDRF1
Genomic Map