siRNA Details
Gene Symbol: ARPC4-TTLL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198793.1 | NP_001185722.1 | 9 | 1595 | ||
GenBank mRNA | BC025289.1 | - | 8 | 1029 |
Gene Symbol: TTLL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025930.3 | NP_001021100.3 | 8 | 1314 | ||
NR_037162.1 | 8 | 1936 | ||||
GenBank mRNA | AK023960.1 | - | 8 | 931 | ||
AK097236.1 | - | 2 | 1552 | |||
AK125875.1 | - | 4 | 3382 | |||
AK297289.1 | - | 5 | 658 | |||
AK315140.1 | - | 6 | 704 | |||
AL096725.1 | - | 2 | 1093 | |||
AL833939.1 | - | 4 | 1741 | |||
BC098298.1 | - | 2 | 561 | 40025661 | ||
BC098361.1 | - | 2 | 561 | |||
BC099735.1 | - | 2 | 561 | |||
BC105638.1 | - | 2 | 561 |
