siRNA Details
| Gene Symbol: RIBC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015653.4 | NP_056468.3 | 3 | 606 | ||
| XM_005261524.4 | XP_005261581.1 | 3 | 431 | |||
| XM_017028766.1 | XP_016884255.1 | 3 | 528 | |||
| GenBank mRNA | AK098586.1 | - | 3 | 606 | ||
| AK225722.1 | - | 3 | 587 | |||
| AL442116.1 | - | 3 | 557 | |||
| BC003024.1 | - | 3 | 568 | 2822570 | ||
| BC009904.2 | - | 3 | 568 | 2822570 | ||
| CR456438.1 | - | 1 | 200 | |||
| CR457398.1 | - | 1 | 191 | |||

RIBC2
Genomic Map