siRNA Details
Gene Symbol: OR1A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012352.2 | NP_036484.1 | 1 | 765 | ||
GenBank mRNA | BC069092.1 | - | 1 | 765 | 7216906 | |
BC106733.1 | - | 1 | 763 | 40015715 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: OR1A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012352.2 | NP_036484.1 | 1 | 765 | ||
GenBank mRNA | BC069092.1 | - | 1 | 765 | 7216906 | |
BC106733.1 | - | 1 | 763 | 40015715 |