siRNA Details
| Gene Symbol: FBXW2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012164.3 | NP_036296.2 | 3 | 668 | ||
| XM_005251910.3 | XP_005251967.1 | 3 | 671 | |||
| XM_005251913.2 | XP_005251970.1 | 3 | 570 | |||
| XM_006717049.3 | XP_006717112.1 | 3 | 853 | |||
| XM_006717051.2 | XP_006717114.1 | 3 | 908 | |||
| XM_011518518.1 | XP_011516820.1 | 3 | 908 | |||
| XM_017014615.1 | XP_016870104.1 | 3 | 671 | |||
| XM_017014616.1 | XP_016870105.1 | 3 | 767 | |||
| XM_017014617.1 | XP_016870106.1 | 3 | 791 | |||
| GenBank mRNA | AF129531.1 | - | 1 | 430 | ||
| AF145024.1 | - | 3 | 575 | |||
| AF176698.1 | - | 2 | 553 | |||
| AF193594.1 | - | 3 | 575 | |||
| AK022484.1 | - | 3 | 618 | |||
| AK300066.1 | - | 3 | 499 | |||
| AK314385.1 | - | 3 | 853 | |||
| BC018738.1 | - | 3 | 584 | 4554961 | ||
| BC110334.1 | - | 2 | 568 | 5741924 | ||

FBXW2
Genomic Map