siRNA Details
| Gene Symbol: OR2F1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012369.2 | NP_036501.2 | 1 | 687 | ||
| XM_011516019.2 | XP_011514321.1 | 4 | 1233 | |||
| XM_011516020.2 | XP_011514322.1 | 4 | 1008 | |||
| GenBank mRNA | BC104975.1 | - | 1 | 687 | 8143978 | |

OR2F1
Genomic Map