siRNA Details
Gene Symbol: OR2B6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012367.1 | NP_036499.1 | 1 | 698 | ||
GenBank mRNA | BC109251.1 | - | 1 | 698 | 40023640 | |
BC110457.2 | - | 1 | 698 | 40022996 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: OR2B6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012367.1 | NP_036499.1 | 1 | 698 | ||
GenBank mRNA | BC109251.1 | - | 1 | 698 | 40023640 | |
BC110457.2 | - | 1 | 698 | 40022996 |