siRNA Details
| Gene Symbol: STAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317769.1 | NP_001304698.1 | 6 | 730 | ||
| NM_012108.3 | NP_036240.1 | 6 | 730 | |||
| XM_017008018.1 | XP_016863507.1 | 6 | 730 | |||
| GenBank mRNA | AB023483.1 | - | 6 | 700 | ||
| AK313676.1 | - | 6 | 679 | |||
| BC014958.1 | - | 6 | 649 | 4851485 | ||

STAP1
Genomic Map