siRNA Details
| Gene Symbol: FBXO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012168.5 | NP_036300.2 | 5 | 1150 | ||
| GenBank mRNA | AF174594.1 | - | 4 | 542 | ||
| AF187318.1 | - | 5 | 998 | |||
| AK308531.1 | - | 6 | 1132 | |||
| AK313019.1 | - | 5 | 750 | |||
| BC025233.1 | - | 5 | 734 | 5090925 | ||
| BC096747.1 | - | 5 | 750 | 5259298 | ||
| CR542110.1 | - | 5 | 659 | |||

FBXO2
Genomic Map