siRNA Details
| Gene Symbol: FBXL6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012162.3 | NP_036294.2 | 8 | 1501 | ||
| NM_024555.5 | NP_078831.4 | 8 | 1483 | |||
| GenBank mRNA | AF370422.1 | - | 7 | 2814 | ||
| AK026541.1 | - | 8 | 1432 | |||
| AK223088.1 | - | 8 | 1432 | |||
| AL713783.1 | - | 4 | 1712 | |||
| BC033066.1 | - | 8 | 1490 | 5516945 | ||

FBXL6
Genomic Map