siRNA Details
| Gene Symbol: FBXL6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012162.3 | NP_036294.2 | 6 | 1008 | ||
| NM_024555.5 | NP_078831.4 | 6 | 990 | |||
| GenBank mRNA | AF174592.1 | - | 6 | 665 | ||
| AF370422.1 | - | 5 | 2321 | |||
| AK026541.1 | - | 6 | 939 | |||
| AK223088.1 | - | 6 | 939 | |||
| AL713783.1 | - | 2 | 1219 | |||
| BC033066.1 | - | 6 | 997 | 5516945 | ||

FBXL6
Genomic Map