siRNA Details
| Gene Symbol: FBXL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001193534.1 | NP_001180463.1 | 5 | 781 | ||
| NM_001193535.1 | NP_001180464.1 | 5 | 733 | |||
| NM_012161.3 | NP_036293.1 | 5 | 784 | |||
| NR_036464.1 | 6 | 888 | ||||
| XM_006713959.3 | XP_006714022.1 | 6 | 880 | |||
| XM_011513831.2 | XP_011512133.1 | 5 | 566 | |||
| XM_011513832.2 | XP_011512134.1 | 5 | 713 | |||
| XM_011513833.2 | XP_011512135.1 | 4 | 524 | |||
| XM_017008019.1 | XP_016863508.1 | 7 | 1651 | |||
| GenBank mRNA | AF142481.1 | - | 5 | 706 | ||
| AF157323.1 | - | 5 | 645 | |||
| AF174591.1 | - | 2 | 191 | |||
| AF176700.1 | - | 5 | 668 | |||
| AF199420.1 | - | 5 | 670 | |||
| AK000153.1 | - | 4 | 494 | |||
| AK295510.1 | - | 5 | 703 | |||
| AK310860.1 | - | 5 | 713 | |||
| AL832808.1 | - | 3 | 420 | |||
| BC012320.1 | - | 6 | 817 | |||
| BC030656.1 | - | 5 | 755 | 4837080 | ||
| BX537957.1 | - | 5 | 679 | |||

FBXL5
Genomic Map