siRNA Details
| Gene Symbol: LCE2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014357.4 | NP_055172.1 | 2 | 445 | ||
| GenBank mRNA | AF005080.1 | - | 2 | 442 | ||
| AF086289.1 | - | 2 | 445 | |||
| BC117235.1 | - | 2 | 436 | |||
| BC117237.1 | - | 2 | 436 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LCE2B| Gene Symbol: LCE2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014357.4 | NP_055172.1 | 2 | 445 | ||
| GenBank mRNA | AF005080.1 | - | 2 | 442 | ||
| AF086289.1 | - | 2 | 445 | |||
| BC117235.1 | - | 2 | 436 | |||
| BC117237.1 | - | 2 | 436 | |||