siRNA Details
| Gene Symbol: CCNF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001323538.1 | NP_001310467.1 | 8 | 865 | ||
| NM_001761.2 | NP_001752.2 | 9 | 969 | |||
| XM_017023822.1 | XP_016879311.1 | 9 | 1129 | |||
| XM_017023823.1 | XP_016879312.1 | 9 | 1129 | |||
| GenBank mRNA | AB208829.1 | - | 8 | 776 | ||
| AK313371.1 | - | 9 | 922 | |||
| BC012349.1 | - | 9 | 893 | 4634116 | ||
| U17105.1 | - | 9 | 894 | |||
| Z36714.1 | - | 9 | 924 | |||

CCNF
Genomic Map