siRNA Details
| Gene Symbol: NKX2-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014360.3 | NP_055175.2 | 1 | 271 | ||
| GenBank mRNA | AK313537.1 | - | 1 | 264 | ||
| BC041090.1 | - | 1 | 238 | 5752253 | ||
| DA438523.1 | - | 1 | 271 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NKX2-8| Gene Symbol: NKX2-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014360.3 | NP_055175.2 | 1 | 271 | ||
| GenBank mRNA | AK313537.1 | - | 1 | 264 | ||
| BC041090.1 | - | 1 | 238 | 5752253 | ||
| DA438523.1 | - | 1 | 271 | |||