siRNA Details
| Gene Symbol: FBXO6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018438.5 | NP_060908.1 | 4 | 662 | ||
| XM_005263448.4 | XP_005263505.1 | 4 | 602 | |||
| XM_005263449.4 | XP_005263506.1 | 4 | 549 | |||
| XM_005263451.4 | XP_005263508.1 | 4 | 602 | |||
| XM_006710571.3 | XP_006710634.1 | 4 | 630 | |||
| XM_011541232.2 | XP_011539534.1 | 4 | 596 | |||
| XM_017001007.1 | XP_016856496.1 | 5 | 664 | |||
| GenBank mRNA | AF129536.1 | - | 4 | 631 | ||
| AF233223.1 | - | 3 | 507 | |||
| AK314989.1 | - | 4 | 662 | |||
| BC020880.1 | - | 4 | 604 | 4614957 | ||

FBXO6
Genomic Map