siRNA Details
Gene Symbol: OR5L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004739.1 | NP_001004739.1 | 1 | 506 | ||
GenBank mRNA | X64990.1 | - | 1 | 132 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: OR5L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004739.1 | NP_001004739.1 | 1 | 506 | ||
GenBank mRNA | X64990.1 | - | 1 | 132 |