siRNA Details
| Gene Symbol: GNL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014366.4 | NP_055181.3 | 12 | 1434 | ||
| NM_206825.1 | NP_996561.1 | 12 | 1531 | |||
| NM_206826.1 | NP_996562.1 | 12 | 1384 | |||
| GenBank mRNA | AF191018.1 | - | 12 | 1431 | ||
| AK027514.1 | - | 12 | 1435 | |||
| AK027516.1 | - | 12 | 1284 | |||
| AK315484.1 | - | 12 | 1347 | |||
| AY825265.1 | - | 11 | 1261 | |||
| BC001024.2 | - | 12 | 1282 | 3342771 | ||

GNL3
Genomic Map