siRNA Details
| Gene Symbol: CNNM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017623.4 | NP_060093.3 | 3 | 1405 | ||
| NM_199078.2 | NP_951060.1 | 2 | 1261 | |||
| XM_011510957.2 | XP_011509259.1 | 3 | 1765 | |||
| XM_017003800.1 | XP_016859289.1 | 3 | 1765 | |||
| GenBank mRNA | AF216965.1 | - | 3 | 504 | ||
| AK000025.1 | - | 2 | 323 | |||
| AK126847.1 | - | 3 | 1405 | |||
| BC007199.1 | - | 2 | 417 | 2964706 | ||
| BC018604.1 | - | 2 | 150 | |||
| BC037272.1 | - | 3 | 1414 | 4823510 | ||
| CR749702.1 | - | 2 | 39 | |||

CNNM3
Genomic Map