siRNA Details
| Gene Symbol: TIMM8B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012459.2 | NP_036591.2 | 2 | 182 | ||
| NR_028383.1 | 3 | 515 | ||||
| GenBank mRNA | AF150087.1 | - | 2 | 119 | ||
| AF152350.1 | - | 2 | 107 | |||
| AF165967.1 | - | 3 | 425 | |||
| AK025397.1 | - | 2 | 140 | |||
| AK312169.1 | - | 2 | 133 | |||
| BC000711.2 | - | 3 | 462 | 2823930 | ||
| BC105986.1 | - | 2 | 150 | 4510312 | ||
| BC106067.1 | - | 2 | 150 | 4708781 | ||

TIMM8B
Genomic Map