siRNA Details
| Gene Symbol: OR11A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013937.3 | NP_039225.1 | 1 | 737 | ||
| GenBank mRNA | AJ459857.1 | - | 1 | 737 | ||
| BC131608.1 | - | 1 | 718 | 40114992 | ||
More Information
Set Membership: |
|

OR11A1
Genomic Map