siRNA Details
| Gene Symbol: OR10G2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005466.2 | NP_001005466.2 | 1 | 949 | ||
| GenBank mRNA | BC137371.1 | - | 1 | 949 | 9021369 | |
| BC137382.1 | - | 1 | 949 | 9021380 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_146271, NM_146431, BC112968, BC148301, BC119377, and others.

OR10G2
Genomic Map