siRNA Details
| Gene Symbol: ITGB1BP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001303277.1 | NP_001290206.1 | 8 | 783 | ||
| NM_012278.2 | NP_036410.1 | 11 | 1036 | |||
| XM_017029405.1 | XP_016884894.1 | 9 | 1306 | |||
| GenBank mRNA | AF110225.1 | - | 11 | 990 | ||
| AF140690.1 | - | 11 | 1006 | |||
| BC108901.2 | - | 10 | 1263 | 40004692 | ||

ITGB1BP2
Genomic Map