siRNA Details
| Gene Symbol: OSTF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012383.4 | NP_036515.4 | 2 | 277 | ||
| XM_006717053.3 | XP_006717116.1 | 2 | 286 | |||
| XM_011518524.2 | XP_011516826.1 | 2 | 67 | |||
| XM_011518525.2 | XP_011516827.1 | 2 | 288 | |||
| XM_017014622.1 | XP_016870111.1 | 2 | 278 | |||
| GenBank mRNA | AK222596.1 | - | 2 | 221 | ||
| AK291511.1 | - | 2 | 245 | |||
| BC007459.1 | - | 2 | 183 | 4052054 | ||
| BP312980.1 | - | 2 | 277 | |||
| U63717.1 | - | 2 | 215 | |||

OSTF1
Genomic Map