siRNA Details
| Gene Symbol: OR8B2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005468.1 | NP_001005468.1 | 1 | 393 | ||
| XM_017017535.1 | XP_016873024.1 | 3 | 625 | |||
| XM_017017536.1 | XP_016873025.1 | 3 | 611 | |||
| Gene Symbol: OR8B3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005467.1 | NP_001005467.1 | 1 | 393 | ||
| XM_017017716.1 | XP_016873205.1 | 6 | 1759 | |||
| XM_017017717.1 | XP_016873206.1 | 6 | 1062 | |||
| XM_017017718.1 | XP_016873207.1 | 4 | 1490 | |||

OR8B2
Genomic Map