siRNA Details
| Gene Symbol: DNAI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001281428.1 | NP_001268357.1 | 14, 15 | 1662 | ||
| NM_012144.3 | NP_036276.1 | 14, 15 | 1650 | |||
| XM_006716758.3 | XP_006716821.1 | 13, 14 | 1293 | |||
| XM_011517846.1 | XP_011516148.1 | 14, 15 | 1662 | |||
| XM_011517847.2 | XP_011516149.1 | 14, 15 | 1662 | |||
| XM_011517849.1 | XP_011516151.1 | 14, 15 | 1662 | |||
| GenBank mRNA | AF091619.1 | - | 14, 15 | 1567 | ||
| AK290696.1 | - | 14, 15 | 1574 | |||
| AK302186.1 | - | 11, 12 | 1193 | |||
| AK303937.1 | - | 11, 12 | 1147 | |||
| AK314639.1 | - | 14, 15 | 1562 | |||
| BC030583.2 | - | 14, 15 | 1578 | 4837991 | ||

DNAI1
Genomic Map