siRNA Details
| Gene Symbol: CD3E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000733.3 | NP_000724.1 | 6 | 499 | ||
| GenBank mRNA | AK292612.1 | - | 6 | 348 | ||
| AK301255.1 | - | 6 | 349 | |||
| AK311641.1 | - | 5 | 796 | |||
| BC049847.1 | - | 5 | 311 | 5214563 | ||
| BI910359.1 | - | 6 | 534 | |||
| BP366535.1 | - | 6 | 516 | |||
| X03884.1 | - | 5 | 297 | |||

CD3E
Genomic Map