siRNA Details
| Gene Symbol: PKD2L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001258448.1 | NP_001245377.1 | 4 | 356 | ||
| NM_001258449.1 | NP_001245378.1 | 4 | 356 | |||
| NM_001300921.1 | NP_001287850.1 | 4 | 356 | |||
| NM_014386.3 | NP_055201.2 | 4 | 356 | |||
| XM_011543318.2 | XP_011541620.1 | 3 | 270 | |||
| XM_011543321.1 | XP_011541623.1 | 4 | 290 | |||
| XM_017009343.1 | XP_016864832.1 | 4 | 372 | |||
| XM_017009344.1 | XP_016864833.1 | 3 | 480 | |||
| XM_017009345.1 | XP_016864834.1 | 4 | 372 | |||
| GenBank mRNA | AF118125.1 | - | 4 | 323 | ||
| AF182034.1 | - | 4 | 326 | |||
| AK301924.1 | - | 4 | 331 | |||
| BC034619.1 | - | 4 | 1409 | |||
| BC038714.1 | - | 4 | 292 | |||
| BC044581.1 | - | 4 | 355 | 4830805 | ||
| BG719682.1 | - | 4 | 320 | |||
| BG721989.1 | - | 4 | 361 | |||

PKD2L2
Genomic Map