siRNA Details
| Gene Symbol: GHITM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014394.2 | NP_055209.2 | 9 | 1174 | ||
| GenBank mRNA | AB009685.1 | - | 8 | 970 | ||
| AF060923.1 | - | 9 | 1048 | |||
| AF078863.1 | - | 9 | 1098 | |||
| AF131820.1 | - | 9 | 1101 | |||
| AK098770.1 | - | 10 | 987 | |||
| AK130094.1 | - | 9 | 1077 | |||
| AK130097.1 | - | 9 | 1042 | |||
| AK225318.1 | - | 9 | 1051 | |||
| AK292864.1 | - | 9 | 1042 | |||
| AK297960.1 | - | 9 | 967 | |||
| AK298332.1 | - | 9 | 1056 | |||
| AK299937.1 | - | 8 | 1080 | |||
| AL136713.1 | - | 9 | 1084 | |||
| AL834502.1 | - | 2 | 75 | |||
| AY358824.1 | - | 9 | 1049 | |||
| BC010354.1 | - | 9 | 1045 | 4093037 | ||
| BC022872.1 | - | 9 | 1056 | |||
| CR533519.1 | - | 8 | 970 | |||

GHITM
Genomic Map