siRNA Details
| Gene Symbol: B9D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243473.2 | NP_001230402.1 | 4 | 375 | ||
| NM_001243475.2 | NP_001230404.1 | 4 | 571 | |||
| NM_001321214.1 | NP_001308143.1 | 4 | 468 | |||
| NM_001321215.1 | NP_001308144.1 | 4 | 468 | |||
| NM_001321216.1 | NP_001308145.1 | 4 | 468 | |||
| NM_001321217.1 | NP_001308146.1 | 4 | 468 | |||
| NM_001321218.1 | NP_001308147.1 | 4 | 468 | |||
| NM_001321219.1 | NP_001308148.1 | 4 | 468 | |||
| NM_015681.4 | NP_056496.1 | 4 | 468 | |||
| XM_005256607.2 | XP_005256664.1 | 4 | 440 | |||
| XM_005256610.1 | XP_005256667.1 | 4 | 443 | |||
| XM_017024452.1 | XP_016879941.1 | 7 | 2832 | |||
| GenBank mRNA | AB030506.1 | - | 4 | 457 | ||
| AK293815.1 | - | 4 | 441 | |||
| AK297782.1 | - | 4 | 358 | |||
| AK309320.1 | - | 4 | 560 | |||
| BC002944.2 | - | 4 | 399 | 3953287 | ||
| BI833202.1 | - | 4 | 463 | |||

B9D1
Genomic Map