siRNA Details
| Gene Symbol: ST6GALNAC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175039.3 | NP_778204.1 | 5 | 979 | ||
| NM_175040.3 | NP_778205.1 | 4 | 892 | |||
| XM_011518527.2 | XP_011516829.1 | 6 | 1071 | |||
| XM_011518529.2 | XP_011516831.1 | 6 | 1102 | |||
| GenBank mRNA | AB035172.1 | - | 4 | 724 | ||
| AJ271734.1 | - | 5 | 994 | |||
| AK000600.1 | - | 5 | 975 | |||
| AK292049.1 | - | 4 | 883 | |||
| BC036705.2 | - | 5 | 885 | 4309208 | ||

ST6GALNAC4
Genomic Map