siRNA Details
| Gene Symbol: ARRDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025604.2 | NP_001020775.1 | 3 | 613 | ||
| NM_001286826.1 | NP_001273755.1 | 3 | 588 | |||
| NM_015683.1 | NP_056498.1 | 3 | 588 | |||
| GenBank mRNA | AF131826.1 | - | 2 | 120 | ||
| AF193051.1 | - | 3 | 588 | |||
| AK000689.1 | - | 3 | 586 | |||
| AK127940.1 | - | 3 | 577 | |||
| AK128588.1 | - | 1 | 905 | |||
| AK314657.1 | - | 3 | 586 | |||
| BC022516.1 | - | 3 | 586 | 4817429 | ||
| BG756314.1 | - | 3 | 615 | |||

ARRDC2
Genomic Map