siRNA Details
| Gene Symbol: ARRDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025604.2 | NP_001020775.1 | 7 | 1292 | ||
| NM_001286826.1 | NP_001273755.1 | 7 | 1267 | |||
| NM_015683.1 | NP_056498.1 | 7 | 1267 | |||
| GenBank mRNA | AF131826.1 | - | 6 | 799 | ||
| AF193051.1 | - | 7 | 1267 | |||
| AK000689.1 | - | 7 | 1265 | |||
| AK127940.1 | - | 7 | 1256 | |||
| AK128588.1 | - | 1 | 2514 | |||
| AK314657.1 | - | 7 | 1265 | |||
| BC022516.1 | - | 7 | 1265 | 4817429 | ||

ARRDC2
Genomic Map