siRNA Details
| Gene Symbol: INPP5J | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002837.2 | NP_001002837.1 | 8 | 972 | ||
| NM_001284285.1 | NP_001271214.1 | 8 | 2076 | |||
| NM_001284286.1 | NP_001271215.1 | 9 | 2198 | |||
| NM_001284287.1 | NP_001271216.1 | 7 | 992 | |||
| NM_001284288.1 | NP_001271217.1 | 9 | 1150 | |||
| NM_001284289.1 | NP_001271218.1 | 10 | 1272 | |||
| XM_011530143.1 | XP_011528445.1 | 10 | 1742 | |||
| XM_017028771.1 | XP_016884260.1 | 10 | 2730 | |||
| XM_017028772.1 | XP_016884261.1 | 9 | 1250 | |||
| XM_017028773.1 | XP_016884262.1 | 10 | 1508 | |||
| GenBank mRNA | AK092859.1 | - | 8 | 2032 | ||
| AK095944.1 | - | 9 | 1079 | |||
| AK293992.1 | - | 9 | 2155 | |||
| AK309548.1 | - | 7 | 931 | |||
| BC109288.2 | - | 10 | 1184 | 40034071 | ||
| U45975.1 | - | 2 | 157 | |||

INPP5J
Genomic Map