siRNA Details
| Gene Symbol: CYTH4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318024.1 | NP_001304953.1 | 11 | 1116 | ||
| NM_013385.4 | NP_037517.1 | 11 | 1097 | |||
| XM_011530147.2 | XP_011528449.1 | 11 | 984 | |||
| XM_011530148.1 | XP_011528450.1 | 12 | 1061 | |||
| GenBank mRNA | AF075458.1 | - | 11 | 1026 | ||
| AF125349.1 | - | 11 | 1097 | |||
| AK002145.1 | - | 5 | 413 | |||
| AK304453.1 | - | 11 | 1017 | |||
| AK315052.1 | - | 11 | 933 | |||
| BC041161.1 | - | 11 | 955 | 5745783 | ||
| CR456551.1 | - | 11 | 946 | |||

CYTH4
Genomic Map