siRNA Details
Gene Symbol: CYTH4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318024.1 | NP_001304953.1 | 8 | 765 | ||
NM_013385.4 | NP_037517.1 | 8 | 746 | |||
XM_011530147.2 | XP_011528449.1 | 8 | 633 | |||
XM_011530148.1 | XP_011528450.1 | 8 | 595 | |||
GenBank mRNA | AF075458.1 | - | 8 | 675 | ||
AF125349.1 | - | 8 | 746 | |||
AK002145.1 | - | 2 | 62 | |||
AK024428.1 | - | 8 | 600 | |||
AK304453.1 | - | 8 | 666 | |||
AK315052.1 | - | 8 | 582 | |||
BC041161.1 | - | 8 | 604 | 5745783 | ||
BP306826.1 | - | 5 | 267 | |||
CR456551.1 | - | 8 | 595 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001112701, NM_011181, BC004662, BC088833, AK168979, and others.
Panther Classification:
Gene Ontology Categories:
