siRNA Details
| Gene Symbol: CPNE7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014427.4 | NP_055242.1 | 9 | 1056 | ||
| NM_153636.2 | NP_705900.1 | 7 | 831 | |||
| XM_011523000.1 | XP_011521302.1 | 7 | 826 | |||
| XM_011523001.2 | XP_011521303.1 | 7 | 591 | |||
| XM_017023138.1 | XP_016878627.1 | 9 | 1020 | |||
| XM_017023139.1 | XP_016878628.1 | 9 | 1031 | |||
| XM_017023140.1 | XP_016878629.1 | 7 | 806 | |||
| XM_017023141.1 | XP_016878630.1 | 6 | 1913 | |||
| GenBank mRNA | AJ133798.1 | - | 9 | 1055 | ||
| AL539371.3 | - | 7 | 833 | |||
| BC035334.1 | - | 7 | 769 | 5191900 | ||
| BC064577.1 | - | 7 | 808 | 5727324 | ||

CPNE7
Genomic Map