siRNA Details
| Gene Symbol: CD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001254750.1 | NP_001241679.1 | 7 | 1447 | ||
| NM_001254751.1 | NP_001241680.1 | 7 | 1447 | |||
| NM_006725.4 | NP_006716.3 | 7 | 1447 | |||
| NR_045638.1 | 7 | 1447 | ||||
| XM_006718738.1 | XP_006718801.1 | 7 | 1447 | |||
| XM_006718739.1 | XP_006718802.1 | 7 | 1447 | |||
| XM_006718740.1 | XP_006718803.1 | 7 | 1447 | |||
| XM_006718741.1 | XP_006718804.1 | 7 | 1447 | |||
| XM_011545360.1 | XP_011543662.1 | 7 | 1447 | |||
| XM_011545362.1 | XP_011543664.1 | 6 | 1144 | |||
| GenBank mRNA | AK128419.1 | - | 8 | 1351 | ||
| AK292704.1 | - | 7 | 1323 | |||
| AK312749.1 | - | 7 | 1408 | |||
| BC033755.1 | - | 7 | 1363 | 5213263 | ||
| BC078669.1 | - | 7 | 1386 | 6302947 | ||
| U66142.1 | - | 7 | 1222 | |||
| U66145.1 | - | 7 | 1222 | |||
| U66146.1 | - | 7 | 1222 | |||
| X60992.1 | - | 7 | 1342 | |||

CD6
Genomic Map