siRNA Details
Gene Symbol: DENND2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB033103.2 | - | 9, 10 | 2312 | ||
AK290056.1 | - | 10, 11 | 2183 | |||
BC132875.1 | - | 8, 9 | 2009 | 40146897 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: DENND2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB033103.2 | - | 9, 10 | 2312 | ||
AK290056.1 | - | 10, 11 | 2183 | |||
BC132875.1 | - | 8, 9 | 2009 | 40146897 |