siRNA Details
| Gene Symbol: STK36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243313.1 | NP_001230242.1 | 3 | 479 | ||
| NM_015690.4 | NP_056505.2 | 3 | 479 | |||
| XM_005246464.1 | XP_005246521.1 | 3 | 367 | |||
| XM_011510959.1 | XP_011509261.1 | 3 | 479 | |||
| XM_011510960.1 | XP_011509262.1 | 3 | 420 | |||
| XM_017003804.1 | XP_016859293.1 | 3 | 367 | |||
| XM_017003805.1 | XP_016859294.1 | 2 | 147 | |||
| GenBank mRNA | AB033104.1 | - | 3 | 436 | ||
| AF200815.1 | - | 2 | 200 | |||
| BC026158.2 | - | 3 | 419 | 4839214 | ||
| DB062910.1 | - | 3 | 481 | |||

STK36
Genomic Map