siRNA Details
| Gene Symbol: CPAMD8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015692.2 | NP_056507.2 | 20, 21 | 2682 | ||
| XM_011527917.1 | XP_011526219.1 | 20, 21 | 2649 | |||
| XM_011527918.1 | XP_011526220.1 | 20, 21 | 2685 | |||
| XM_011527919.1 | XP_011526221.1 | 20, 21 | 2685 | |||
| XM_011527920.2 | XP_011526222.1 | 20, 21 | 2685 | |||
| XM_011527921.2 | XP_011526223.1 | 20, 21 | 2685 | |||
| XM_011527922.1 | XP_011526224.1 | 20, 21 | 2685 | |||
| XM_011527923.1 | XP_011526225.1 | 20, 21 | 2685 | |||
| XM_011527924.2 | XP_011526226.1 | 20, 21 | 2685 | |||
| XM_011527925.1 | XP_011526227.1 | 4, 5 | 527 | |||
| XM_017026594.1 | XP_016882083.1 | 20, 21 | 2685 | |||
| GenBank mRNA | AB033109.1 | - | 20, 21 | 2507 | ||
| AY101765.1 | - | 20, 21 | 2526 | |||

CPAMD8
Genomic Map