siRNA Details
| Gene Symbol: RSPH14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014433.2 | NP_055248.1 | 6, 7 | 942 | ||
| GenBank mRNA | AF133587.1 | - | 6, 7 | 942 | ||
| BC008986.1 | - | 6, 7 | 910 | 4339021 | ||
| CR456567.1 | - | 5, 6 | 842 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RSPH14| Gene Symbol: RSPH14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014433.2 | NP_055248.1 | 6, 7 | 942 | ||
| GenBank mRNA | AF133587.1 | - | 6, 7 | 942 | ||
| BC008986.1 | - | 6, 7 | 910 | 4339021 | ||
| CR456567.1 | - | 5, 6 | 842 | |||