siRNA Details
| Gene Symbol: NAAA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042402.1 | NP_001035861.1 | 3 | 544 | ||
| NM_014435.3 | NP_055250.2 | 3 | 544 | |||
| XM_005262920.3 | XP_005262977.1 | 3 | 542 | |||
| XM_005262921.3 | XP_005262978.1 | 3 | 542 | |||
| XM_005262923.2 | XP_005262980.1 | 3 | 542 | |||
| XM_005262924.2 | XP_005262981.1 | 3 | 542 | |||
| XM_005262925.3 | XP_005262982.1 | 3 | 542 | |||
| XM_006714180.2 | XP_006714243.1 | 3 | 542 | |||
| XM_011531852.2 | XP_011530154.1 | 3 | 542 | |||
| XM_011531854.2 | XP_011530156.1 | 3 | 542 | |||
| XM_017008027.1 | XP_016863516.1 | 3 | 542 | |||
| XM_017008028.1 | XP_016863517.1 | 3 | 542 | |||
| XM_017008029.1 | XP_016863518.1 | 3 | 542 | |||
| XM_017008030.1 | XP_016863519.1 | 3 | 542 | |||
| GenBank mRNA | AB161353.1 | - | 3 | 480 | ||
| AK024677.1 | - | 1044 | ||||
| AK301132.1 | - | 2 | 369 | |||
| AK309271.1 | - | 3 | 516 | |||
| BC006388.2 | - | 3 | 524 | 4122755 | ||
| BM763257.1 | - | 3 | 544 | |||
| M92449.1 | - | 2 | 240 | |||

NAAA
Genomic Map