siRNA Details
| Gene Symbol: SLC39A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271957.1 | NP_001258886.1 | 4 | 1196 | ||
| NM_001271958.1 | NP_001258887.1 | 4 | 1033 | |||
| NM_001271959.1 | NP_001258888.1 | 4 | 1202 | |||
| NM_001271960.1 | NP_001258889.1 | 5 | 1094 | |||
| NM_001271961.1 | NP_001258890.1 | 3 | 902 | |||
| NM_014437.4 | NP_055252.2 | 5 | 1340 | |||
| GenBank mRNA | AF132942.1 | - | 3 | 890 | ||
| AF151829.1 | - | 4 | 911 | |||
| AJ243649.1 | - | 4 | 927 | |||
| AJ271671.1 | - | 4 | 941 | |||
| AK074943.1 | - | 4 | 1150 | |||
| AK075257.1 | - | 4 | 964 | |||
| AK293389.1 | - | 3 | 810 | |||
| AK297983.1 | - | 5 | 908 | |||
| AK300244.1 | - | 5 | 900 | |||
| BC002563.2 | - | 4 | 915 | 3138575 | ||
| BC003152.2 | - | 3 | 887 | 2958651 | ||
| BC007886.2 | - | 4 | 912 | 4122086 | ||
| BC014303.2 | - | 5 | 1320 | 3942586 | ||
| CR457217.1 | - | 3 | 854 | |||

SLC39A1
Genomic Map