siRNA Details
| Gene Symbol: TSNAX-DISC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028393.1 | 6 | 1335 | |||
| NR_028394.1 | 7 | 1463 | ||||
| NR_028395.1 | 7 | 1463 | ||||
| NR_028396.1 | 6 | 1335 | ||||
| NR_028397.1 | 6 | 1200 | ||||
| NR_028398.1 | 4 | 954 | ||||
| NR_028399.1 | 8 | 1570 | ||||
| NR_028400.1 | 7 | 1442 | ||||

DISC1
Genomic Map