siRNA Details
| Gene Symbol: IL17C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013278.3 | NP_037410.1 | 3 | 447 | ||
| GenBank mRNA | AF152099.1 | - | 3 | 447 | ||
| AY358471.1 | - | 3 | 447 | |||
| BC069152.1 | - | 2 | 398 | 7217012 | ||
| BC101835.1 | - | 3 | 442 | 8069341 | ||
| BC112196.1 | - | 3 | 442 | 8327664 | ||

IL17C
Genomic Map