siRNA Details
| Gene Symbol: IL17B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317987.1 | NP_001304916.1 | 3 | 689 | ||
| NM_014443.2 | NP_055258.1 | 3 | 665 | |||
| XM_011537622.2 | XP_011535924.1 | 4 | 988 | |||
| XM_017009346.1 | XP_016864835.1 | 3 | 1182 | |||
| XM_017009347.1 | XP_016864836.1 | 5 | 896 | |||
| XM_017009348.1 | XP_016864837.1 | 4 | 779 | |||
| GenBank mRNA | AF110385.1 | - | 3 | 658 | ||
| AF152098.1 | - | 3 | 656 | |||
| AF184969.1 | - | 3 | 656 | |||
| AF212311.1 | - | 3 | 665 | |||
| AF218727.1 | - | 3 | 656 | |||
| AY358931.1 | - | 3 | 656 | |||

IL17B
Genomic Map